1-(1H-Imidazol-4-ylmethyl)piperidine-4-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:914637-46-0
:
Formula:C₁₀H₁₅N₃O₂
MDL Number:MFCD08235217
:209.25 g/mol
Storage Temperature:Ambient
Frequently Bought Together