C2 Ceramide
Supplier: Apollo Scientific
Jump to:
About this item
A biologically active, cell permeable, non-physiological ceramide analog. It inhibits cell proliferation, induces monocytic differentiation of HL-60 cells & inducesapoptosis.
- CAS Number:3102-57-6
- Formula:C₂₀H₃₉NO₃
- MDL Number:MFCD00153903
- Molecular Weight:341.536 g/mol
- Boiling Pt:87…89 °C
- Storage Temperature:Freezer
2 Options Available Below
Product Details & Documents
A biologically active, cell permeable, non-physiological ceramide analog. It inhibits cell proliferation, induces monocytic differentiation of HL-60 cells & inducesapoptosis.
- CAS Number:3102-57-6
- Formula:C₂₀H₃₉NO₃
- MDL Number:MFCD00153903
- Molecular Weight:341.536 g/mol
- Boiling Pt:87…89 °C
- Storage Temperature:Freezer
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



