Valeric acid 99%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Pentanoic acid
- CAS Number:109-52-4
- Formula:C₅H₁₀O₂
- MDL Number:MFCD00004413
- Molecular Weight:102.13 g/mol
- Boiling Pt:184…186 °C
- Melting Pt:–34 °C
- Density:0.938 g/cm³ (25 °C)
- Storage Temperature:Ambient
- UN:3265
- ADR:8,III
2 Options Available Below
Product Details & Documents
Synonyms: Pentanoic acid
- CAS Number:109-52-4
- Formula:C₅H₁₀O₂
- MDL Number:MFCD00004413
- Molecular Weight:102.13 g/mol
- Boiling Pt:184…186 °C
- Melting Pt:–34 °C
- Density:0.938 g/cm³ (25 °C)
- Storage Temperature:Ambient
- UN:3265
- ADR:8,III



