Order Entry
Denmark
ContactUsLinkComponent
2-Methylene-1,3-propanediol 95%
Irritant
2-Methylene-1,3-propanediol 95%
Catalog # APOSOR54371-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Methylene-1,3-propanediol 95%
Catalog # APOSOR54371-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :88.11 g/mol
Boiling Pt:93…95 °C/2mmHg
Density:1.081 g/cm³ (25 °C)
Flash Pt:110 °C
EINECS:222-516-7
 :GHS07
Storage Temperature:Ambient
CAS Number:3513-81-3
Formula:C₄H₈O₂
MDL Number:MFCD00075162