
3,5-Dihydroxytoluene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:504-15-4
:
Formula:C₇H₈O₂
MDL Number:MFCD00002291
:124.14 g/mol
Boiling Pt:287…290 °C
Melting Pt:106…112 °C
Density:1.29 g/cm³ (25 °C)
Flash Pt:159 °C
EINECS:207-984-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together