Order Entry
Denmark
ContactUsLinkComponent
N-Carbobenzoxy-DL-serine 98%
N-Carbobenzoxy-DL-serine 98%
Catalog # APOSOR471643-100G
Supplier:  Apollo Scientific
CAS Number:  
N-Carbobenzoxy-DL-serine 98%
Catalog # APOSOR471643-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :239.23 g/mol
Storage Temperature:Ambient
CAS Number:2768-56-1
Formula:C₁₁H₁₃NO₅
MDL Number:MFCD00063143