1-Chloropropane
Catalog # APOSOR460042-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:78.54 g/mol
Boiling Pt:46…47 °C
Melting Pt:–123 °C
Density:0.892 g/cm³ (25 °C)
Flash Pt:–18 °C
UN:1278
ADR:3,II
:GHS02
:GHS07
Storage Temperature:Refrigerator
CAS Number:540-54-5
Formula:C₃H₇Cl
MDL Number:MFCD00000995