
cis-tert-Butyl-N-(3-aminocyclobutyl)carbamate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1212395-34-0
:
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD11099896
:186.26 g/mol
Melting Pt:111…112 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together