

α-Bromo-3-iodotoulene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:49617-83-6
:
Formula:C₇H₆BrI
MDL Number:MFCD00019016
:296.93 g/mol
Melting Pt:45…50 °C
Flash Pt:113 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
:GHS05
:GHS07
Frequently Bought Together