
5-Aminobenzene-1,3-diol hydrochloride
Catalog # APOSOR22059-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:161.5862 g/mol
Melting Pt:300 °C
:GHS07
Storage Temperature:Ambient
CAS Number:6318-56-5
Formula:C₆H₈ClNO₂
MDL Number:MFCD00662897