Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
4-Methylmorpholin

4-Methylmorpholin

Flammable
Corrosive
Irritant
Supplier:   Apollo Scientific
4-Methylmorpholin
Supplier:  Apollo Scientific
Certificates
4-Methylmorpholin

2 Options Available Below

Product Details & Documents
Synonyms:  NMM, N-Methylmorpholine
  • CAS Number:109-02-4
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD00006175
  • Molecular Weight:101.15 g/mol
  • Boiling Pt:115…116 °C
  • Melting Pt:–66 °C
  • Density:0,92 g/cm³ (25 °C)
  • Flash Pt:14 °C
  • Storage Temperature:Stue temperatur
  • EINECS:203-640-0
  • UN:2535
  • ADR:3+8,II
Product Family Options

Product Information

  • Pakning
    Pakn
    Availability
    Price
  • Bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - kr231.00
    kr231.00
    1 * 100 mL
       kr231.00

  • Bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr571.00
    kr571.00
    1 * 500 mL
       kr571.00

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.