Baicalein
Supplier: Apollo Scientific
Jump to:
About this item
An inhibitor of 12-lipoxygenase, leukotriene biosynthesis & release of lysosomal enzymes. It also inhibits cellular calcium-ion uptake, mobilization & adjuvant-induced arthirritis.
Synonyms: 5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one
- CAS Number:491-67-8
- Formula:C₁₅H₁₀O₅
- MDL Number:MFCD00017459
- Molecular Weight:270.24 g/mol
- Melting Pt:264…265 °C
- Storage Temperature:Refrigerator
2 Options Available Below
Product Details & Documents
An inhibitor of 12-lipoxygenase, leukotriene biosynthesis & release of lysosomal enzymes. It also inhibits cellular calcium-ion uptake, mobilization & adjuvant-induced arthirritis.
Synonyms: 5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one
- CAS Number:491-67-8
- Formula:C₁₅H₁₀O₅
- MDL Number:MFCD00017459
- Molecular Weight:270.24 g/mol
- Melting Pt:264…265 °C
- Storage Temperature:Refrigerator



