Order Entry
Denmark
ContactUsLinkComponent
2-Butyne-1-ol 97%
Flammable
Irritant
2-Butyne-1-ol 97%
Catalog # APOSOR14505-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
2-Butyne-1-ol 97%
Catalog # APOSOR14505-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :70.09 g/mol
Boiling Pt:142…143 °C
Melting Pt:–2 °C
Density:0.938 g/cm³ (25 °C)
Flash Pt:51 °C
Storage Temperature:Ambient
EINECS:212-113-4
UN:1987
ADR:3,III
 :GHS02
 :GHS07
CAS Number:764-01-2
Formula:C₄H₆O
MDL Number:MFCD00002914