
Pyrrolidin-2-ylmethanol
Catalog # APOSOR13968-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pakning:Bottle
- Pakn:25 g
- Seq:3308
Specifications
About this item
:
:101.15 g/mol
Boiling Pt:76 °C/ 0,2mmHg
Density:1,04 g/cm³ (25 °C)
:GHS07
Storage Temperature:Stue temperatur
CAS Number:498-63-5
Formula:C5H11NO
MDL Number:MFCD00601073