Order Entry
Denmark
Orders LinkContactUsLinkComponent
3-Phenoxybenzyl bromide

3-Phenoxybenzyl bromide

Corrosive
Irritant
New Product
Supplier:   Apollo Scientific
3-Phenoxybenzyl bromide
Supplier:  Apollo Scientific
New Product
Certificates
3-Phenoxybenzyl bromide

4 Options Available Below

Product Details & Documents
Synonyms:  1-(Bromomethyl)-3-phenoxybenzene, 3-(Bromomethyl)diphenyl ether
  • CAS Number:51632-16-7
  • Formula:C₁₃H₁₁BrO
  • MDL Number:MFCD01761658
  • Molecular Weight:263.13 g/mol
  • Boiling Pt:112…118 °C/0.1mmHg
  • Storage Temperature:Ambient
  • EINECS:257-327-9
  • UN:3265
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 472,00 kr
    472,00 kr
    1 * 250 mg
       472,00 kr

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 758,00 kr
    758,00 kr
    1 * 1 g
       758,00 kr

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 2.360,00 kr
    2.360,00 kr
    1 * 5 g
       2.360,00 kr

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 6.350,00 kr
    6.350,00 kr
    1 * 25 g
       6.350,00 kr

    Specifications:
    New Product
    New Product