Order Entry
Denmark
Orders LinkContactUsLinkComponent
Vanillic acid

Vanillic acid

Irritant
Supplier:   Apollo Scientific
Vanillic acid
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzoic acid
  • CAS Number:121-34-6
  • Formula:C₈H₈O₄
  • MDL Number:MFCD00002551
  • Molecular Weight:168.15 g/mol
  • Melting Pt:210…213 °C
  • Storage Temperature:Ambient
  • EINECS:204-466-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 165,00 kr
    165,00 kr
    1 * 5 g
       165,00 kr

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 187,00 kr
    187,00 kr
    1 * 25 g
       187,00 kr

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 330,00 kr
    330,00 kr
    1 * 100 g
       330,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.