AMPA
Catalog # APOSOR1150T-10MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:10 mg
- Seq:2275
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3-(3-Hydroxy-5-methylisoxazol-4-yl)-DL-alanine
:
:186.2 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:77521-29-0
Formula:C₇H₁₀N₂O₄
Melting Pt:>200 °C (dec.)