Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Luminol (3-Aminophthalhydrazide)
Luminol (3-Aminophthalhydrazide) 98%
Luminol (3-Aminophthalhydrazide) 98%
APOSOR11118-25G
:
Apollo Scientific
:
Luminol (3-Aminophthalhydrazide) 98%
APOSOR11118-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
319…320 °C
:
177.16 g/mol
:
208-309-4
:
GHS07
:
Ambient
:
521-31-3
:
C₈H₇N₃O₂
:
MFCD00006890