
4-Bromo-2-chlorophenol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3964-56-5
:
Formula:C₆H₄BrClO
MDL Number:MFCD00002166
:207.45 g/mol
Boiling Pt:232…235 °C
Melting Pt:47…49 °C
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together