Order Entry
Denmark
ContactUsLinkComponent
Vanadium(II) chloride 85%
Vanadium(II) chloride 85%
Catalog # APOSIN3760-5G
Supplier:  Apollo Scientific
CAS Number:  
Vanadium(II) chloride 85%
Catalog # APOSIN3760-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :121.85 g/mol
Melting Pt:910 °C (subl.)
Density:3.09 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:234-176-7
UN:3260
ADR:8,II
 :GHS05
 :GHS07
CAS Number:10580-52-6
Formula:Cl₂V