Potassium dihydrogen phosphate 99,9%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Melting Pt:252…253 °C
Density:2.338 g/cm³ (25 °C)
EINECS:231-913-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together