
Magnesium oxide 99.99%
:1309-48-4
:
:MgO
:MFCD00011109
:40.3 g/mol
:3600 °C
:2800 °C
:3.58 g/cm³ (25 °C)
:GHS07
:Ambient
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Magnesium oxide ≥99% (metals basis), nanopowder
: Thermo Fisher Scientific