Spermidine
Supplier: Apollo Scientific
Jump to:
About this item
Used in the isolation of chromosomes.
Synonyms: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
- CAS Number:124-20-9
- Formula:C₇H₁₉N₃
- MDL Number:MFCD00008229
- Molecular Weight:145.25 g/mol
- Melting Pt:24 °C
- Density:0.925 g/cm³ (25 °C)
- Flash Pt:112 °C
- Storage Temperature:Refrigerator
- UN:2735
- ADR:8,II
2 Options Available Below
Product Details & Documents
Used in the isolation of chromosomes.
Synonyms: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
- CAS Number:124-20-9
- Formula:C₇H₁₉N₃
- MDL Number:MFCD00008229
- Molecular Weight:145.25 g/mol
- Melting Pt:24 °C
- Density:0.925 g/cm³ (25 °C)
- Flash Pt:112 °C
- Storage Temperature:Refrigerator
- UN:2735
- ADR:8,II
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



