Order Entry
Denmark
Orders LinkContactUsLinkComponent
Biotin-PEG11-NH2

Biotin-PEG11-NH2

Bottle
Catalog # APOSBIPG1213-1G
New Product
Supplier:   Apollo Scientific
Biotin-PEG11-NH2
Catalog # APOSBIPG1213-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  604786-74-5
Biotin-PEG11-NH2

Bottle

CAS Number:  604786-74-5
Price
7.080,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Biotin-PEG11-Amine
  • CAS Number:604786-74-5
  • Formula:C₃₄H₆₆N₄O₁₃S
  • Molecular Weight:770.97 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.