

Phorbol-12-myristate-13-acetate ≥99.5%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16561-29-8
:
Formula:C₃₆H₅₆O₈
MDL Number:MFCD00036736
:616.836 g/mol
Melting Pt:50…70 °C
:GHS07
:GHS08
Storage Temperature:Freezer
Frequently Bought Together