
Minocycline hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13614-98-7
:
Formula:C₂₃H₂₇N₃O₇
MDL Number:MFCD00083669
:493.94 g/mol
EINECS:237-099-7
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together