
Resveratrol
Catalog # APOSBIK9013-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:228.25 g/mol
Melting Pt:261…267 °C
:GHS07
Storage Temperature:Freezer
CAS Number:501-36-0
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00133799