Hydroxyurea 98%
Supplier: Apollo Scientific
Jump to:
About this item
Anti-neoplastic. Inactivates ribonucleoside reductase byformation of free radical nitroxide. This blocks synthesis of deoxynucleotides which inhibits DNA synthesis.
- Protein
Synonyms: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- CAS Number:127-07-1
- Formula:CH₄N₂O₂
- MDL Number:MFCD00007943
- Molecular Weight:76.05 g/mol
- Melting Pt:135 °C
- Storage Temperature:Refrigerator
- EINECS:204-821-7
3 Options Available Below
Product Details & Documents
Anti-neoplastic. Inactivates ribonucleoside reductase byformation of free radical nitroxide. This blocks synthesis of deoxynucleotides which inhibits DNA synthesis.
- Protein
Synonyms: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- CAS Number:127-07-1
- Formula:CH₄N₂O₂
- MDL Number:MFCD00007943
- Molecular Weight:76.05 g/mol
- Melting Pt:135 °C
- Storage Temperature:Refrigerator
- EINECS:204-821-7
Product Family Options
Product Information
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



