β-Nicotinamide mononucleotide
:1094-61-7
:
:C₁₁H₁₅N₂O₈P
:MFCD00038748
:334.22 g/mol
:166 °C (dec.)
:Freezer
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
VWR® Disposable Microcentrifuge Tubes
: VWR Collection
Transfer pipettes
: RATIOLAB
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Premium Line, Serological Pipettes
: VWR Collection
Measuring scoops
: VITLAB
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Acetonitrile ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection