Order Entry
Denmark
Orders LinkContactUsLinkComponent
DIPSO ≥98%

DIPSO ≥98%

New Product
Supplier:   Apollo Scientific
DIPSO ≥98%
Supplier:  Apollo Scientific
New Product
Certificates

4 Options Available Below

Product Details & Documents

Biological buffer - useful range 7.0 - 8.2.

US Patent No. 4,169,950 (To Research Organics)

Synonyms:  3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
  • CAS Number:68399-80-4
  • Formula:C₇H₁₇NO₆S
  • MDL Number:MFCD00038353
  • Molecular Weight:261.29 g/mol
  • Melting Pt:189…192 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 165,00 kr
    165,00 kr
    1 * 5 g
       165,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 187,00 kr
    187,00 kr
    1 * 25 g
       187,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 4.560,00 kr
    4.560,00 kr
    1 * 2,5 kg
       4.560,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 17.760,00 kr
    17.760,00 kr
    1 * 10 kg
       17.760,00 kr

    Specifications:
    New Product
    New Product