Biotin hydrazide
Leverandør: Apollo Scientific
Jump to:
About this item
Carbohydrate reactive. Microscopy studies on membranesRef: O'Shannessy,D.J.et al. (1984) Immun.Letters 8,273-277
Synonyms: 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide
- CAS Number:66640-86-6
- Formula:C₁₀H₁₈N₄O₂S
- MDL Number:MFCD00078532
- Molecular Weight:258.34 g/mol
- Melting Pt:240…244 °C
- Storage Temperature:Refrigerator
5 Options Available Below
Product Details & Documents
Carbohydrate reactive. Microscopy studies on membranesRef: O'Shannessy,D.J.et al. (1984) Immun.Letters 8,273-277
Synonyms: 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide
- CAS Number:66640-86-6
- Formula:C₁₀H₁₈N₄O₂S
- MDL Number:MFCD00078532
- Molecular Weight:258.34 g/mol
- Melting Pt:240…244 °C
- Storage Temperature:Refrigerator
Product Family Options
Product Information
- Pack typePkTilgængelighedPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



