
Alcian blue 8GX
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:33864-99-2
:75881-23-1
Formula:C₅₆H₆₈Cl₄CuN₁₆S₄
MDL Number:MFCD00010720
:1298.86 g/mol
Melting Pt:148 °C
EINECS:278-333-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together