
(R)-tert-Butyl-2-aminopropanoate hydrochloride
Catalog # APOSBIA0105-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:181.7 g/mol
Melting Pt:170…175 °C
:GHS07
Storage Temperature:Ambient
CAS Number:59531-86-1
Formula:C₇H₁₅NO₂·HCl
MDL Number:MFCD00077087