About this item
Premium 150 and Premium 90 HR colony counters and zone measurements systems.
- Plates can be automatically read at the press of a button
- High definition colour images taken with a 1,4 MP scientific grade CCD camera ensure that even the smallest colony can be seen and counted, while zone measurements are fast, accurate and reproducible
- A unique 3-channel LED lighting system configured for exceptional illumination of all sample types
- Can be used with a new or existing stand-alone desktop or laptop computers via USB connection – no additional PCI cards are required
The Premium 150 accepts plates of up to 150 mm in diameter and detect colonies as small as 100 µm and measure zones accurately to 0,5 mm. Designed for use across a wide range of applications.
The Premium 90 HR is equipped with a high resolution 12 mm F1.4 lens, accept plates of up to 90 mm and detect colonies as small as 75 µm and measure zones accurately to 0,5 mm.
Suitable for a wide range of applications in a pharma environment.
The Premium 150 and Premium 90 HR include colony counting and inhibition zone measuring ability, spiral plate counting, Ames testing and multi-sector measurements. Other applications on demand. Results can be automatically transferred to Excel, PDF or a LIMS system. All data generated is GLP/GMP/21 CFR Part 11 compliant with a full audit trail and can be used to produce professional reports. Each system comes complete with two validation plates.
Specifications
- Catalog No:
- 710-2074
- 710-2073
- Count:
- Separation of touching colonies, exclusion areas, colour mode, shape mode, size mode
- Separation of touching colonies, exclusion areas, colour mode, shape mode, size mode
- Illumination:
- Unique 3-channel (red, green, blue) LED lighting, multi-array LED lighting (computer controlled), lower lighting with upper reflective lighting for all applications
- Unique 3-channel (red, green, blue) LED lighting, multi-array LED lighting (computer controlled), lower lighting with upper reflective lighting for all applications
- Camera:
- Integrated, sensitive 1,4 MP
- Integrated, sensitive 1,4 MP
- Software:
- Windows 7/10 compatible
- Windows 7/10 compatible
- W×D×H:
- 400×500×450 mm
- 400×500×450 mm
- Weight:
- 20 kg
- 20 kg
Specifications
Related Information
Automatic Colony Counters, Premium 150 and Premium 90 HR.mp4 Demo Automatic Colony Counters, Premium 150 and Premium 90 HR
Videos
Manual Quick Guide VWR Colony Counter Premium Full Installation Guide Manual Quick Guide VWR Colony Counter Premium Inhibition Zone Batch set up Manual Quick Guide VWR Colony Counter Premium Total Plate Count Manual VWR Colony Counter Premium EN 200521 Quick Guide VWR Colony Counter Premium Chromogenic ID
Manual
VWR Colony counters