5-(Trifluoromethyl)benzene-1,3-diol 95%
Catalog # APOSPC910295-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:178.109 g/mol
Storage Temperature:Ambient
CAS Number:454-68-2
Formula:C₇H₅F₃O₂
MDL Number:MFCD00061289