Order Entry
Denmark
Orders LinkContactUsLinkComponent
2H-Tridecafluoro-2-methylpentane 97%

2H-Tridecafluoro-2-methylpentane 97%

Irritant
Supplier:   Apollo Scientific
2H-Tridecafluoro-2-methylpentane 97%
Supplier:  Apollo Scientific
Certificates
2H-Tridecafluoro-2-methylpentane 97%

3 Options Available Below

Product Details & Documents
Synonyms:  1,1,1,2,2,3,3,5,5,5-Decafluoro-4-(trifluoromethyl)pentane
  • CAS Number:30320-28-6
  • Formula:C₆HF₁₃
  • MDL Number:MFCD09763637
  • Molecular Weight:320.051 g/mol
  • Boiling Pt:60…61 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 845,00 kr
    845,00 kr
    1 * 1 g
       845,00 kr

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 2.520,00 kr
    2.520,00 kr
    1 * 5 g
       2.520,00 kr

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 6.370,00 kr
    6.370,00 kr
    1 * 25 g
       6.370,00 kr

    Specifications:
    New Product
    New Product