
1H,1H,2H,2H-Perfluorooctylmethyldimethoxysilane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85857-17-6
:
Formula:C₁₁H₁₃F₁₃O₂Si
MDL Number:MFCD00153260
:452.284 g/mol
Boiling Pt:80 °C (9 mm Hg)
Density:1.434 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together