
1H,1H,1H,2H,2H-Nonafluorohexane 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:38436-17-8
:
Formula:C₆H₅F₉
MDL Number:MFCD07784187
:248.09 g/mol
Boiling Pt:63 °C
Density:1.428 g/cm³ (25 °C)
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together