Order Entry
Denmark
ContactUsLinkComponent
1H,1H,2H,2H-Perfluorohexyl isopropyl carbonate
1H,1H,2H,2H-Perfluorohexyl isopropyl carbonate
Catalog # APOSPC450440-25G
Supplier:  Apollo Scientific
CAS Number:  
1H,1H,2H,2H-Perfluorohexyl isopropyl carbonate
Catalog # APOSPC450440-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :350.178 g/mol
Storage Temperature:Ambient
CAS Number:1980086-72-3
Formula:C₁₀H₁₁F₉O₃
MDL Number:MFCD28334415