1H,1H,2H,2H-Perfluorohexyl propyl carbonate
Catalog # APOSPC450427-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:350.178 g/mol
Storage Temperature:Ambient
CAS Number:1980034-59-0
Formula:C₁₀H₁₁F₉O₃
MDL Number:MFCD28334403