Order Entry
Denmark
ContactUsLinkComponent
1H,1H,9H-Perfluorononyl 1H,1H,2H,2H-perfluorohexyl carbonate
1H,1H,9H-Perfluorononyl 1H,1H,2H,2H-perfluorohexyl carbonate
Catalog # APOSPC450380-25G
Supplier:  Apollo Scientific
CAS Number:  
1H,1H,9H-Perfluorononyl 1H,1H,2H,2H-perfluorohexyl carbonate
Catalog # APOSPC450380-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :722.185 g/mol
Storage Temperature:Ambient
CAS Number:1980039-76-6
Formula:C₁₆H₇F₂₅O₃
MDL Number:MFCD28334372