Order Entry
Denmark
ContactUsLinkComponent
2-Amino-2-methyl-3,3,3-trifluoropropanoic acid, N-CBZ protected 97%
Irritant
2-Amino-2-methyl-3,3,3-trifluoropropanoic acid, N-CBZ protected 97%
Catalog # APOSPC2175-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Amino-2-methyl-3,3,3-trifluoropropanoic acid, N-CBZ protected 97%
Catalog # APOSPC2175-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :291.223 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:139520-43-7
Formula:C₁₂H₁₂F₃NO₄
MDL Number:MFCD07777230