3-Methoxycyclobutane-1-carboxylic acid
Catalog # APOSOR922058-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:130.142 g/mol
Storage Temperature:Ambient
CAS Number:480450-03-1
Formula:C₆H₁₀O₃
MDL Number:MFCD08705860