N'-[3-(3-Aminopropylamino)propyl]butane-1,4-diamine hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1255099-40-1
:
Formula:C₁₀H₂₆N₄·ClH
MDL Number:MFCD17926313
Molecular Weight:238.801 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together