Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Cbz-3-methyl-DL-valine methyl ester
N-Cbz-3-methyl-DL-valine methyl ester
N-Cbz-3-methyl-DL-valine methyl ester
N-Cbz-3-methyl-DL-valine methyl ester
APOSOR471304-1G
:
Apollo Scientific
:
N-Cbz-3-methyl-DL-valine methyl ester
APOSOR471304-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
279.332 g/mol
:
Ambient
:
141971-09-7
:
C₁₅H₂₁NO₄
:
MFCD11113181