Order Entry
Denmark
ContactUsLinkComponent
(S)-a-(Fmoc-amino)cyclobutaneacetic acid 95%
(S)-a-(Fmoc-amino)cyclobutaneacetic acid 95%
Catalog # APOSOR451176-250MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-a-(Fmoc-amino)cyclobutaneacetic acid 95%
Catalog # APOSOR451176-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :337.369 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1391630-31-1
Formula:C₂₀H₁₉NO₄
MDL Number:MFCD09839305