2-Amino-2-cyclopropylethan-1-ol hydrochloride
Catalog # APOSOR451109-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:137.608 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1306603-98-4
Formula:C₅H₁₁NO·ClH
MDL Number:MFCD18839092