Methyl (2Z)-3-(dimethylamino)-2-(phenylformamido)prop-2-enoate 95%
Catalog # APOSOR32527-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:248.278 g/mol
Storage Temperature:Ambient
CAS Number:125008-68-6
Formula:C₁₃H₁₆N₂O₃
MDL Number:MFCD00202513