(1R)-1-(2-methoxyphenyl)-2,2-dimethylpropane-1,3-diol
Catalog # APOSOR311296-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:210.27 g/mol
Storage Temperature:Ambient
CAS Number:167255-26-7
Formula:C₁₂H₁₈O₃
MDL Number:MFCD28976646