Bis(4-(4-propylcyclohexyl)aniline) sulphuric acid
Catalog # APOSOR310784-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:532.79 g/mol
Storage Temperature:Ambient
CAS Number:1980039-16-4
Formula:C₃₀H₄₈N₂O₄S
MDL Number:MFCD28023536